Assay Details
| Gene Symbol: Acsm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144748.1 | NP_653349.1 | 3 - 4 | 489 | 99 | |
| XM_008759677.1 | XP_008757899.1 | 3 - 4 | 474 | 99 | ||
| XM_008759678.1 | XP_008757900.1 | 3 - 4 | 511 | 99 | ||
| XM_008759679.1 | XP_008757901.1 | 3 - 4 | 512 | 99 | ||
| GenBank mRNA | AF062389.1 | - | 3 - 4 | 489 | 99 | |
| BC078721.1 | - | 3 - 4 | 466 | 7110246 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsm2a
Genomic Map