This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00690138_m1
See other ACADM GE Assays ›
Gene Symbol:
Acadm
Gene Name:
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases:
MCAD
Chromosome Location:
Chr.2: 260124418 - 260148589 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00690138_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acadm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016986.2 NP_058682.2 8 - 9 822 78
GenBank mRNA FQ214755.1 - 2 - 3 150 78
J02791.1 - 8 - 9 737 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200004932.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acadm

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

liver development
glycogen biosynthetic process
organic acid metabolic process
regulation of gluconeogenesis
fatty acid beta-oxidation
heart development
response to nutrient
response to cold
carnitine metabolic process
response to hormone
post-embryonic development
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to drug
response to starvation
carnitine biosynthetic process
response to copper ion
protein homotetramerization
response to glucocorticoid
medium-chain fatty acid metabolic process
medium-chain fatty acid catabolic process
cardiac muscle cell differentiation
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
isomerase activity
identical protein binding
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200034127.rn.1 TC0200004932.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0