This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00690751_g1
See other GLUL GE Assays ›
Gene Symbol:
Glul
Gene Name:
glutamate-ammonia ligase
Gene Aliases:
Glns
Chromosome Location:
Chr.13: 71331052 - 71340207 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00690751_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glul
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017073.3 NP_058769.4 6 - 7 921 90
GenBank mRNA BC061559.1 - 6 - 7 934 6918514 90
BC072694.1 - 6 - 7 921 7103110 90
FQ213256.1 - 6 - 7 932 90
FQ216093.1 - 6 - 7 928 90
FQ226911.1 - 6 - 7 944 90
FQ227474.1 - 6 - 7 935 90
FQ233770.1 - 6 - 7 935 90
M29579.1 - 6 - 7 932 90
M91652.1 - 6 - 7 930 90
X07921.1 - 6 - 7 932 90

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glul

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

glutamate metabolic process
glutamine biosynthetic process
cell proliferation
cellular response to starvation
response to glucose
ammonia assimilation cycle
positive regulation of insulin secretion
neurotransmitter metabolic process
positive regulation of epithelial cell proliferation
protein homooligomerization
positive regulation of synaptic transmission, glutamatergic
magnesium ion binding
glutamate decarboxylase activity
glutamate-ammonia ligase activity
ATP binding
glutamate binding
manganese ion binding
identical protein binding
dynein light chain binding

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P381069 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P381069 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P381069 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P381069 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1030714 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P458704 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P381069 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1386870_at - -
Rat Genome 230 2.0 Array 1367633_PM_at - -
Rat Genome 230 2.0 Array 1367633_at - -
Rat Genome 230 2.0 Array 1386870_PM_at - -
Rat Genome 230 2.0 Array 1367632_at - -
Rat Genome 230 2.0 Array 1367632_PM_at - -
Affymetrix HT Rat Focus Array 1367633_at - -
Affymetrix HT Rat Focus Array 1367633_PM_at - -
Affymetrix HT Rat Focus Array 1367632_at - -
Affymetrix HT Rat Focus Array 1367632_PM_at - -
Affymetrix HT Rat Focus Array 1386870_PM_at - -
Affymetrix HT Rat Focus Array 1386870_at - -
Affymetrix Rat Clariom S Assay PSR1300003566.rn.1 TC1300000506.rn.1 RTA TC
Rat Expression Set 230 1367633_PM_at - -
Rat Expression Set 230 1386870_PM_at - -
Rat Expression Set 230 1367633_at - -
Rat Expression Set 230 1386870_at - -
Rat Expression Set 230 1367632_at - -
Rat Expression Set 230 1367632_PM_at - -
Rat Genome U34 Set rc_AA852004_s_at - -
Rat Genome U34 Set rc_AI232783_s_at - -
Rat Genome U34 Set rc_AI012265_i_at - -
Rat Genome U34 Set rc_AI232783_s_at - -
Rat Genome U34 Set rc_AI012265_i_at - -
Rat Genome U34 Set rc_AA852004_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20788710 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0