This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00690933_m1
See other PPIA GE Assays ›
Gene Symbol:
Ppia
Gene Name:
peptidylprolyl isomerase A (cyclophilin A)
Gene Aliases:
CYCA, CyP-A
Chromosome Location:
Chr.14: 86673775 - 86677443 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00690933_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017101.1 NP_058797.1 3 - 4 233 149
GenBank mRNA BC059141.1 - 3 - 4 238 6919594 149
BC091153.1 - 3 - 4 209 7308693 149
FQ210268.1 - 3 - 4 234 149
FQ222826.1 - 3 - 4 234 149
FQ223330.1 - 3 - 4 235 149
FQ223366.1 - 3 - 4 234 149
FQ223500.1 - 3 - 4 234 149
FQ227889.1 - 3 - 4 234 149
FQ228411.1 - 3 - 4 234 149
FQ231674.1 - 3 - 4 234 149
FQ233894.1 - 3 - 4 233 149
FQ234872.1 - 3 - 4 234 149
M19533.1 - 3 - 4 233 149
M25637.1 - 3 - 4 226 149

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Endogenous Controls Recommended Assay Rattus norvegicus Rat Chemokine Panel Rat Endogenous Controls Inventoried Rat_Endogenous_Controls Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppia

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
neuron differentiation
negative regulation of protein phosphatase type 2B activity
lipid particle organization
regulation of viral genome replication
positive regulation of viral genome replication
positive regulation of protein secretion
peptidyl-prolyl cis-trans isomerase activity
cyclosporin A binding
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P016970 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058353 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058351 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 40090 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 916363 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 21608 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016970 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058353 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058351 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016970 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058351 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058353 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058353 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P016970 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058351 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P595960 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700509919 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 702451356 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 37696 - -
Agilent Rat cDNA Microarray (G4105A) 700509919 - -
Agilent Rat cDNA Microarray (G4105A) 702451356 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10771987 - -
Affymetrix Rat Gene 1.0 ST Array 10867046 - -
Affymetrix Rat Gene 1.0 ST Array 10774096 - -
Affymetrix Rat Gene 1.0 ST Array 10771985 - -
Affymetrix Rat Gene 1.0 ST Array 10774095 - -
Affymetrix Rat Gene 1.0 ST Array 10774099 - -
Affymetrix Rat Gene 1.0 ST Array 10774097 - -
Affymetrix Rat Gene 1.0 ST Array 10791473 - -
Affymetrix Rat Gene 1.0 ST Array 10853756 - -
Affymetrix Rat Gene 1.0 ST Array 10771986 - -
Affymetrix Rat Gene 1.0 ST Array 10774098 - -
Affymetrix Rat Gene 1.1 ST Array 10867045 - -
Affymetrix Rat Gene 1.1 ST Array 10771984 - -
Affymetrix Rat Gene 1.1 ST Array 10774094 - -
Affymetrix Rat Gene 1.1 ST Array 10853755 - -
Affymetrix Rat Gene 1.1 ST Array 10791472 - -
Rat Genome 230 2.0 Array 1398850_PM_at - -
Rat Genome 230 2.0 Array 1398850_at - -
Affymetrix HT Rat Focus Array 1398850_at - -
Affymetrix HT Rat Focus Array 1398850_PM_at - -
Affymetrix Rat Clariom S Assay PSR1400007082.rn.1 TC1400000951.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400007078.rn.1 TC1400000951.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400002594.rn.1 TC1400000315.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400007071.rn.1 TC1400000951.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400007076.rn.1 TC1400000951.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400002593.rn.1 TC1400000315.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400007080.rn.1 TC1400000951.rn.1 RTA TC
Rat Expression Set 230 1398850_PM_at - -
Rat Expression Set 230 1398850_at - -
Affymetrix Rat Gene 1.0 ST Array 10774094 - -
Affymetrix Rat Gene 1.0 ST Array 10791472 - -
Affymetrix Rat Gene 1.0 ST Array 10771984 - -
Affymetrix Rat Gene 1.0 ST Array 10853755 - -
Affymetrix Rat Gene 1.0 ST Array 10867045 - -
Rat Genome U34 Set M19533mRNA_i_at - -
Rat Genome U34 Set rc_AA818152_f_at - -
Rat Genome U34 Set rc_AI228674_s_at - -
Rat Genome U34 Set rc_AA818858_s_at - -
Rat Genome U34 Set rc_AA818858_s_at - -
Rat Genome U34 Set rc_AA818152_f_at - -
Rat Genome U34 Set rc_AI228674_s_at - -
Rat Genome U34 Set M19533mRNA_i_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1372734 - -
RatRef-12 Expression BeadChip ILMN_51793 - -

Back To Top
 

Citations (1)

Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
Authors
Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
Journal
J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 UTC 2009)
Volume (Year)
201 (2009)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline