This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00691397_g1
See other UNC119 GE Assays ›
Gene Symbol:
Unc119
Gene Name:
UNC-119 homolog (C. elegans)
Gene Aliases:
RRG4, Uncl19
Chromosome Location:
Chr.10: 65606919 - 65612318 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00691397_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc119
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017188.1 NP_058884.1 4 - 5 641 87
GenBank mRNA BC062057.1 - 4 - 5 667 5598983 87
U40999.1 - 4 - 5 641 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000001162.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc119

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
visual perception
lipoprotein transport
positive regulation of protein tyrosine kinase activity
negative regulation of clathrin-mediated endocytosis
negative regulation of caveolin-mediated endocytosis
protein binding
lipid binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P134079 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 398679 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 914076 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 432486 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 696201 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134079 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134079 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P134079 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P617594 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700546731 - -
Agilent Rat cDNA Microarray (G4105A) 700546731 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10745075 - -
Affymetrix Rat Gene 1.0 ST Array 10745079 - -
Affymetrix Rat Gene 1.0 ST Array 10745080 - -
Affymetrix Rat Gene 1.0 ST Array 10745077 - -
Affymetrix Rat Gene 1.0 ST Array 10745076 - -
Affymetrix Rat Gene 1.0 ST Array 10745078 - -
Affymetrix Rat Gene 1.1 ST Array 10745074 - -
Rat Genome 230 2.0 Array 1367738_at - -
Rat Genome 230 2.0 Array 1367738_PM_at - -
Affymetrix HT Rat Focus Array 1367738_PM_at - -
Affymetrix HT Rat Focus Array 1367738_at - -
Affymetrix Rat Clariom S Assay PSR1000010640.rn.1 TC1000001162.rn.1 RTA TC
Rat Expression Set 230 1367738_at - -
Rat Expression Set 230 1367738_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10745074 - -
Rat Genome U34 Set U40999_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22232678 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1372394 - -
RatRef-12 Expression BeadChip ILMN_66815 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0