This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00691438_g1
See other SSR4 GE Assays ›
Gene Symbol:
Ssr4
Gene Name:
signal sequence receptor, delta
Gene Aliases:
-
Chromosome Location:
Chr.X: 156995763 - 156999709 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00691438_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017199.1 NP_058895.1 3 - 4 421 63
XM_006229571.2 XP_006229633.1 3 - 4 477 63
GenBank mRNA BC058482.1 - 3 - 4 334 6921620 63
FQ221421.1 - 1 - 2 37 63
FQ221423.1 - 1 - 2 37 63
FQ221949.1 - 1 - 2 37 63
FQ228311.1 - 1 - 2 37 63
FQ228501.1 - 1 - 2 37 63
Z19087.1 - 3 - 4 421 63

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr4

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00016466.rn.1 TC0X00002644.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0