This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00692187_m1
See other BAAT GE Assays ›
Gene Symbol:
Baat
Gene Name:
bile acid CoA:amino acid N-acyltransferase
Gene Aliases:
kan-1
Chromosome Location:
Chr.5: 64768397 - 64777368 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00692187_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Baat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017300.2 NP_058996.2 3 - 4 862 112
GenBank mRNA BC088153.1 - 3 - 4 862 7368634 112
D43964.1 - 3 - 4 877 112
FQ218773.1 - 906 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500002714.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Baat

Panther Classification:

Molecular Function -

esterase hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

liver development
bile acid conjugation
glycine metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
bile acid biosynthetic process
bile acid metabolic process
taurine metabolic process
organ regeneration
molecular_function
receptor binding
palmitoyl-CoA hydrolase activity
N-acyltransferase activity
glycine N-choloyltransferase activity
carboxylic ester hydrolase activity
medium-chain acyl-CoA hydrolase activity
long-chain acyl-CoA hydrolase activity
very long chain acyl-CoA hydrolase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119369 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 828014 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 80272 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 251954 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 680890 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 156889 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 675216 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 920310 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 180255 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 166867 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119369 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119369 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119369 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421685 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700481559 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 14102 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 36425 - -
Agilent Rat cDNA Microarray (G4105A) 700481559 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10876713 - -
Affymetrix Rat Gene 1.0 ST Array 10876712 - -
Affymetrix Rat Gene 1.0 ST Array 10876716 - -
Affymetrix Rat Gene 1.0 ST Array 10876714 - -
Affymetrix Rat Gene 1.0 ST Array 10876715 - -
Affymetrix Rat Gene 1.1 ST Array 10876711 - -
Rat Genome 230 2.0 Array 1387508_PM_at - -
Rat Genome 230 2.0 Array 1387508_at - -
Affymetrix HT Rat Focus Array 1387508_at - -
Affymetrix HT Rat Focus Array 1387508_PM_at - -
Affymetrix Rat Clariom S Assay PSR0500020257.rn.1 TC0500002714.rn.1 RTA TC
Rat Expression Set 230 1387508_at - -
Rat Expression Set 230 1387508_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10876711 - -
Rat Genome U34 Set D43964_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21982711 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1355867 - -
RatRef-12 Expression BeadChip ILMN_56365 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0