This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00692259_g1
See other ATP5G1 GE Assays ›
Gene Symbol:
Atp5g1
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9)
Gene Aliases:
-
Chromosome Location:
Chr.10: 83895941 - 83898640 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00692259_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5g1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017311.1 NP_059007.1 3 - 4 173 93
XM_006247188.2 XP_006247250.1 3 - 4 216 93
XM_006247189.1 XP_006247251.1 3 - 4 321 93
GenBank mRNA D13123.1 - 3 - 4 173 93
FQ209790.1 - 3 - 4 191 93
FQ210883.1 - 2 - 3 137 93
FQ216886.1 - 3 - 4 191 93
FQ217997.1 - 3 - 4 187 93

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5g1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
response to ethanol
ATP metabolic process
molecular_function
lipid binding
hydrogen ion transmembrane transporter activity

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P110659 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007574 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007574 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007574 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000032528.rn.1 TC1000003650.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0