This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00710093_s1
See other GPBAR1 GE Assays ›
Gene Symbol:
Gpbar1
Gene Name:
G protein-coupled bile acid receptor 1
Gene Aliases:
Tgr5
Chromosome Location:
Chr.9: 81557426 - 81560956 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00710093_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpbar1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177936.1 NP_808797.1 1 944 62
XM_006245235.2 XP_006245297.1 2 1956 62
GenBank mRNA AB086172.1 - 1 944 62
AB089310.1 - 1 944 62
JX534229.1 - 2 1251 62

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpbar1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
bile acid receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141329 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557172 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557172 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P557172 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557172 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10924259 - -
Affymetrix Rat Gene 1.1 ST Array 10924258 - -
Affymetrix Rat Gene 1.0 ST Array 10924258 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21186072 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1362439 - -
RatRef-12 Expression BeadChip ILMN_51323 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline