This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00756347_g1
See other ATP5O GE Assays ›
Gene Symbol:
Atp5o
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Gene Aliases:
Atpo, Oscp
Chromosome Location:
Chr.11: 32081606 - 32087918 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00756347_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5o
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138883.1 NP_620238.1 4 - 5 329 84
GenBank mRNA BC060544.1 - 4 - 5 337 6918775 84
D13127.1 - 4 - 5 329 84
FQ209940.1 - 4 - 5 337 84
FQ210807.1 - 4 - 5 343 84
FQ217675.1 - 4 - 5 334 84
FQ223905.1 - 4 - 5 366 84
FQ226488.1 - 4 - 5 330 84

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5o

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP synthesis coupled proton transport
mitochondrial ATP synthesis coupled proton transport
ATP metabolic process
steroid binding
drug binding
ATPase activity
protein complex binding
proton-transporting ATP synthase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100007850.rn.1 TC1100001317.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0