This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00820752_gH
See other LDHA GE Assays ›
Gene Symbol:
Ldha
Gene Name:
lactate dehydrogenase A
Gene Aliases:
Ldh1
Chromosome Location:
Chr.1: 102900288 - 102909713 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00820752_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ldha
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017025.1 NP_058721.1 4 - 5 522 59
XM_006229232.2 XP_006229294.1 4 - 5 690 59
GenBank mRNA BC060587.1 - 4 - 5 474 6919582 59
BC084698.1 - 3 - 4 424 7122339 59
BC168737.1 - 4 - 5 521 6917937 59
FQ214854.1 - 4 - 5 522 59
X01964.1 - 4 - 5 522 59

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ldha

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
lactate metabolic process
response to nutrient
response to glucose
response to organic cyclic compound
NAD metabolic process
substantia nigra development
cellular response to extracellular stimulus
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
response to estrogen
cellular carbohydrate metabolic process
post-embryonic organ development
response to cAMP
oxidation-reduction process
lactate dehydrogenase activity
L-lactate dehydrogenase activity
protein binding
kinase binding
identical protein binding
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100012365.rn.1 TC0100001528.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0