This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00821401_g1
See other DBI GE Assays ›
Gene Symbol:
Dbi
Gene Name:
diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)
Gene Aliases:
Acbp, Acoabp3, Ep, Odn, RNACOABP3, Ttn
Chromosome Location:
Chr.13: 36147667 - 36156076 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00821401_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dbi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031853.4 NP_114054.1 1 - 2 122 69
GenBank mRNA FQ209835.1 - 1 - 2 70 69
FQ209938.1 - 1 - 2 65 69
FQ210140.1 - 1 - 2 70 69
FQ210165.1 - 1 - 2 65 69
FQ210336.1 - 1 - 2 67 69
FQ210460.1 - 1 - 2 70 69
FQ210781.1 - 1 - 2 97 69
FQ210997.1 - 1 - 2 67 69
FQ211081.1 - 1 - 2 70 69
FQ211192.1 - 1 - 2 70 69
FQ211339.1 - 1 - 2 71 69
FQ213527.1 - 1 - 2 71 69
FQ214572.1 - 1 - 2 70 69
FQ217714.1 - 1 - 2 71 69
FQ218148.1 - 1 - 2 65 69
FQ218164.1 - 1 - 2 89 69
FQ218239.1 - 1 - 2 70 69
FQ218300.1 - 1 - 2 70 69
FQ218680.1 - 1 - 2 70 69
FQ219136.1 - 1 - 2 70 69
FQ219739.1 - 1 - 2 70 69
FQ221153.1 - 1 - 2 65 69
FQ221871.1 - 1 - 2 71 69
FQ222052.1 - 1 - 2 70 69
FQ222378.1 - 1 - 2 72 69
FQ222416.1 - 1 - 2 65 69
FQ222564.1 - 1 - 2 71 69
FQ222579.1 - 1 - 2 101 69
FQ223413.1 - 1 - 2 70 69
FQ223492.1 - 1 - 2 71 69
FQ224361.1 - 1 - 2 70 69
FQ224797.1 - 1 - 2 71 69
FQ229114.1 - 1 - 2 65 69
M14201.1 - 1 - 2 122 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the M14201.1,NM_031853.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dbi

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

behavioral fear response
hair follicle development
acyl-CoA metabolic process
triglyceride metabolic process
steroid biosynthetic process
brain development
learning or memory
glial cell proliferation
protein palmitoylation
lateral ventricle development
pancreatic juice secretion
cellular response to nutrient
negative regulation of fatty acid beta-oxidation
regulation of synaptic transmission, GABAergic
phosphatidylcholine acyl-chain remodeling
skin development
positive regulation of lipid biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
long-term synaptic potentiation
fatty-acyl-CoA binding
protein binding
lipid binding
benzodiazepine receptor binding
long-chain fatty acyl-CoA binding
protein dimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300008444.rn.1 TC1300001270.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300008448.rn.1 TC1300001270.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22265198 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0