This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00821778_g1
See other DCTPP1 GE Assays ›
Gene Symbol:
Dctpp1
Gene Name:
dCTP pyrophosphatase 1
Gene Aliases:
RS21-C6, Rs21c6, Xtp3tpa
Chromosome Location:
Chr.1: 198703459 - 198706852 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00821778_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dctpp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138892.1 NP_620247.1 1 - 2 151 76
GenBank mRNA AF331839.1 - 1 - 2 125 76
AY029335.1 - 1 - 2 151 76

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dctpp1

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleoside triphosphate catabolic process
protein homotetramerization
magnesium ion binding
pyrophosphatase activity
pyrimidine deoxyribonucleotide binding
nucleoside-triphosphate diphosphatase activity
dCTP diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100052840.rn.1 TC0100006695.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100052841.rn.1 TC0100006695.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0