This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00821779_g1
See other DCTPP1 GE Assays ›
Gene Symbol:
Dctpp1
Gene Name:
dCTP pyrophosphatase 1
Gene Aliases:
RS21-C6, Rs21c6, Xtp3tpa
Chromosome Location:
Chr.1: 198703459 - 198706852 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00821779_g1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dctpp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138892.1 NP_620247.1 2 - 3 266 103
GenBank mRNA AF331839.1 - 2 - 3 240 103
AY029335.1 - 2 - 3 266 103

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dctpp1

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleoside triphosphate catabolic process
protein homotetramerization
magnesium ion binding
pyrophosphatase activity
pyrimidine deoxyribonucleotide binding
nucleoside-triphosphate diphosphatase activity
dCTP diphosphatase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P285803 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 111951 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 183185 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P285803 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P285803 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P285803 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P285803 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10789734 - -
Affymetrix Rat Gene 1.0 ST Array 10725991 - -
Affymetrix Rat Gene 1.0 ST Array 10725990 - -
Affymetrix Rat Gene 1.0 ST Array 10725992 - -
Affymetrix Rat Gene 1.1 ST Array 10789732 - -
Affymetrix Rat Gene 1.1 ST Array 10725989 - -
Rat Genome 230 2.0 Array 1370308_PM_at - -
Rat Genome 230 2.0 Array 1370308_at - -
Affymetrix HT Rat Focus Array 1370308_at - -
Affymetrix HT Rat Focus Array 1370308_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100052839.rn.1 TC0100006695.rn.1 RTA TC
Rat Expression Set 230 1370308_at - -
Rat Expression Set 230 1370308_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10725989 - -
Affymetrix Rat Gene 1.0 ST Array 10789732 - -
Rat Genome U34 Set rc_AI101475_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22263767 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360240 - -
RatRef-12 Expression BeadChip ILMN_55996 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0