This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01141061_g1
See other UBE2L3 GE Assays ›
Gene Symbol:
Ube2l3 NEWGENE_1308361
Gene Name:
ubiquitin-conjugating enzyme E2L 3
ubiquitin-conjugating enzyme E2L 3
Gene Aliases:
-
Ube2l3
Chromosome Location:
Chr.11: 88047832 - 88088476 on Build Rnor_6.0
Chromosome Location:
Chr.11: 88047191 - 88086544 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01141061_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108847.1 NP_001102317.1 1 - 2 58 76
Gene Symbol: NEWGENE_1308361
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008768927.1 XP_008767149.1 2 - 3 285 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001108847.1,XM_008768927.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
regulation of transcription, DNA-templated
metabolic process
cell proliferation
protein ubiquitination
positive regulation of protein ubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
cell cycle phase transition
positive regulation of ubiquitin-protein transferase activity
protein K11-linked ubiquitination
cellular response to steroid hormone stimulus
cellular response to glucocorticoid stimulus
transcription coactivator activity
ubiquitin-protein transferase activity
ligase activity
ubiquitin protein ligase binding
poly(A) RNA binding
ubiquitin conjugating enzyme activity
ubiquitin-protein transferase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100006098.rn.1 TC1100001021.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0