This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01158820_m1
See other MAPRE2 GE Assays ›
Gene Symbol:
Mapre2
Gene Name:
microtubule-associated protein, RP/EB family, member 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 14814150 - 15780290 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01158820_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapre2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101000.1 NP_001094470.1 147 72
GenBank mRNA BC105879.1 - 147 7123955 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800000129.rn.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic nuclear division
cell division
microtubule binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047724 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047724 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047724 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047724 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1049020 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1049015 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1372472_at - -
Rat Genome 230 2.0 Array 1372472_PM_at - -
Affymetrix Rat Clariom S Assay PSR1800000891.rn.1 TC1800000129.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800000895.rn.1 TC1800000129.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800000896.rn.1 TC1800000129.rn.1 RTA TC
Rat Expression Set 230 1372472_at - -
Rat Expression Set 230 1372472_PM_at - -
Rat Genome U34 Set rc_AI176863_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1530524 - -
RatRef-12 Expression BeadChip ILMN_160776 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline