This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01185107_g1
See other VAMP1 GE Assays ›
Gene Symbol:
Vamp1
Gene Name:
vesicle-associated membrane protein 1
Gene Aliases:
Syb1
Chromosome Location:
Chr.4: 157726740 - 157733644 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01185107_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vamp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013090.2 NP_037222.2 3 - 4 421 75
GenBank mRNA AF498262.1 - 2 - 3 287 75
BC092206.1 - 3 - 4 319 7313343 75
CB582258.1 - 3 - 4 421 75
EF653274.1 - 3 - 4 346 75
EF653275.1 - 3 - 4 346 75
M24104.1 - 3 - 4 384 75
U74621.1 - 3 - 4 319 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vamp1

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
exocytosis
vesicle fusion
neurotransmitter secretion
vesicle-mediated transport
SNARE complex assembly
SNARE binding
SNAP receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0