This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01258495_m1
See other AFAP1L1 GE Assays ›
Gene Symbol:
Afap1l1
Gene Name:
actin filament associated protein 1-like 1
Gene Aliases:
RGD1311580
Chromosome Location:
Chr.18: 57052662 - 57114580 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01258495_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Afap1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106142.1 NP_001099612.1 12 - 13 1574 66
XM_006254794.2 XP_006254856.1 12 - 13 1609 66
GenBank mRNA FQ211638.1 - 12 - 13 1539 66
FQ231300.1 - 1592 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800001532.rn.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P139932 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P766165 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 473226 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 503830 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 853489 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 484112 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 181319 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 894957 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 573336 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 26304 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 943636 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 868740 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 331445 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 648174 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 355146 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 393853 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 891303 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 771892 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 487137 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 13725 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 587249 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 604236 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 682610 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 485798 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 555888 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 150851 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 858882 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 862415 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84393 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 540980 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 411423 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 776744 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 508263 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 481820 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 162592 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 463652 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 836580 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740026 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 890916 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 523791 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 944617 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 124110 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 142322 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36909 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 273380 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 171109 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 87477 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 153909 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 157754 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 23524 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 74027 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 39703 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 103240 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 119158 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139932 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P766165 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139932 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P766165 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139932 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P766165 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1006717 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 92954 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10804729 - -
Affymetrix Rat Gene 1.0 ST Array 10804743 - -
Affymetrix Rat Gene 1.0 ST Array 10804728 - -
Affymetrix Rat Gene 1.0 ST Array 10804740 - -
Affymetrix Rat Gene 1.0 ST Array 10804738 - -
Affymetrix Rat Gene 1.0 ST Array 10804742 - -
Affymetrix Rat Gene 1.0 ST Array 10804745 - -
Affymetrix Rat Gene 1.0 ST Array 10804746 - -
Affymetrix Rat Gene 1.0 ST Array 10804741 - -
Affymetrix Rat Gene 1.0 ST Array 10804734 - -
Affymetrix Rat Gene 1.0 ST Array 10804730 - -
Affymetrix Rat Gene 1.0 ST Array 10804732 - -
Affymetrix Rat Gene 1.0 ST Array 10804744 - -
Affymetrix Rat Gene 1.0 ST Array 10804726 - -
Affymetrix Rat Gene 1.0 ST Array 10804727 - -
Affymetrix Rat Gene 1.0 ST Array 10804731 - -
Affymetrix Rat Gene 1.0 ST Array 10804747 - -
Affymetrix Rat Gene 1.0 ST Array 10804733 - -
Affymetrix Rat Gene 1.1 ST Array 10804721 - -
Rat Genome 230 2.0 Array 1373181_PM_at - -
Rat Genome 230 2.0 Array 1392482_at - -
Rat Genome 230 2.0 Array 1373181_at - -
Rat Genome 230 2.0 Array 1392482_PM_at - -
Rat Genome 230 2.0 Array 1377658_PM_at - -
Rat Genome 230 2.0 Array 1398206_at - -
Rat Genome 230 2.0 Array 1377658_at - -
Rat Genome 230 2.0 Array 1398206_PM_at - -
Affymetrix HT Rat Focus Array 1373181_PM_at - -
Affymetrix HT Rat Focus Array 1373181_at - -
Affymetrix HT Rat Focus Array 1377658_PM_at - -
Affymetrix HT Rat Focus Array 1377658_at - -
Affymetrix Rat Clariom S Assay PSR1800010843.rn.1 TC1800001532.rn.1 RTA TC
Rat Expression Set 230 1373181_PM_at - -
Rat Expression Set 230 1373181_at - -
Rat Expression Set 230 1377658_PM_at - -
Rat Expression Set 230 1377658_at - -
Affymetrix Rat Gene 1.0 ST Array 10804721 - -
Rat Genome U34 Set rc_AI170431_at - -
Rat Genome U34 Set rc_AI104848_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22124563 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0