This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01289124_m1
See other YTHDF3 GE Assays ›
Gene Symbol:
Ythdf3
Gene Name:
YTH N(6)-methyladenosine RNA binding protein 3
Gene Aliases:
-
Chromosome Location:
Chr.2: 100677388 - 100711365 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01289124_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ythdf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108546.1 NP_001102016.1 3 - 4 371 151
XM_006232168.1 XP_006232230.1 4 - 5 540 297
XM_008760873.1 XP_008759095.1 4 - 5 540 297
XM_008760875.1 XP_008759097.1 3 - 4 362 151

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006232168.1, XM_008760873.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Process(es)

poly(A) RNA binding
N6-methyladenosine-containing RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200005025.rn.1 TC0200000819.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200005028.rn.1 TC0200000819.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0