This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01331653_m1
See other BRF1 GE Assays ›
Gene Symbol:
Brf1
Gene Name:
BRF1, RNA polymerase III transcription initiation factor 90 subunit
Gene Aliases:
-
Chromosome Location:
Chr.6: 137762230 - 137808346 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01331653_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Brf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106761.1 NP_001100231.1 14 - 15 1590 60
XM_006240658.2 XP_006240720.1 14 - 15 1603 60
XM_006240659.2 XP_006240721.2 13 - 14 2700 60
XM_008764929.1 XP_008763151.1 14 - 15 1600 60
XM_008764930.1 XP_008763152.1 12 - 13 1424 60
XM_008764931.1 XP_008763153.1 11 - 12 2429 60
XM_008764932.1 XP_008763154.1 11 - 12 1238 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600003261.rn.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription initiation from RNA polymerase III promoter
translational initiation
regulation of mRNA stability
positive regulation of transcription from RNA polymerase III promoter
molecular_function
translation initiation factor activity
zinc ion binding
TBP-class protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600023038.rn.1 TC0600003261.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0