This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01400495_m1
See other TMBIM1 GE Assays ›
Gene Symbol:
Tmbim1
Gene Name:
transmembrane BAX inhibitor motif containing 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 81569289 - 81586469 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01400495_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmbim1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007713.1 NP_001007714.1 9 - 10 789 66
XM_008767238.1 XP_008765460.1 9 - 10 3161 66
XM_008767239.1 XP_008765461.1 10 - 11 812 66
GenBank mRNA BC079087.1 - 9 - 10 789 7123233 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900002718.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmbim1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
negative regulation of establishment of protein localization to plasma membrane
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of Fas signaling pathway
positive regulation of blood vessel remodeling
death receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0900022413.rn.1 TC0900002718.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0