This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01401244_m1
See other IKZF1 GE Assays ›
Gene Symbol:
Ikzf1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
RGD1562979
Chromosome Location:
Chr.14: 91782270 - 91867828 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01401244_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ikzf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107237.1 NP_001100707.1 2 - 3 249 71
XM_006251491.2 XP_006251553.1 2 - 3 764 71
XM_006251492.2 XP_006251554.1 2 - 3 4599 71
XM_006251493.2 XP_006251555.1 2 - 3 302 71
XM_006251494.2 XP_006251556.2 2 - 3 589 71
XM_006251495.2 XP_006251557.1 2 - 3 277 71
XM_006251496.2 XP_006251558.1 2 - 3 76 71
XM_006251497.2 XP_006251559.1 2 - 3 310 71
XM_008770312.1 XP_008768534.1 2 - 3 589 71
XM_008770313.1 XP_008768535.1 2 - 3 589 71
XM_008770314.1 XP_008768536.1 2 - 3 589 71
XM_008770315.1 XP_008768537.1 2 - 3 589 71
XM_008770316.1 XP_008768538.1 2 - 3 589 71
XM_008770317.1 XP_008768539.1 2 - 3 589 71
XM_008770318.1 XP_008768540.1 2 - 3 589 71
XM_008770319.1 XP_008768541.1 2 - 3 589 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400001003.rn.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
natural killer cell differentiation
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
hemopoiesis
B cell differentiation
T cell differentiation
forebrain development
positive regulation of multicellular organism growth
positive regulation of B cell differentiation
positive regulation of neutrophil differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
lymph node development
thymus development
Peyer's patch development
gland development
positive regulation of NK T cell differentiation
retina development in camera-type eye
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400007413.rn.1 TC1400001003.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0