This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01403644_g1
See other NDUFAF3 GE Assays ›
Gene Symbol:
Ndufaf3
Gene Name:
NADH dehydrogenase (ubiquinone) complex I, assembly factor 3
Gene Aliases:
RGD708545
Chromosome Location:
Chr.8: 117351628 - 117355416 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01403644_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufaf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033971.1 NP_001029143.1 2 - 3 940 89
NM_020080.2 NP_064465.1 2 - 3 958 89
XM_006243845.2 XP_006243907.1 3 - 4 2440 89
GenBank mRNA BC079306.1 - 2 - 3 692 7134644 89
FQ216095.1 - 2 - 3 309 89
FQ233128.1 - 1 - 2 385 89
U95160.1 - 2 - 3 294 89

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufaf3

Gene Ontology Categories:

Function(s)

mitochondrial respiratory chain complex I assembly

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800027492.rn.1 TC0800003380.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0