This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01403708_m1
See other TFDP2 GE Assays ›
Gene Symbol:
Tfdp2
Gene Name:
transcription factor Dp-2 (E2F dimerization partner 2)
Gene Aliases:
Tcfdp2
Chromosome Location:
Chr.8: 104040796 - 104181228 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01403708_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tfdp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106847.2 NP_001100317.2 7 - 8 792 74
XM_006243556.2 XP_006243618.1 1096 74
XM_006243558.2 XP_006243620.1 7 - 8 2195 74
XM_006243561.2 XP_006243623.1 6 - 7 2155 74
XM_006243563.1 XP_006243625.1 9 - 10 1204 74
XM_006243566.2 XP_006243628.1 1397 74
XM_006243567.2 XP_006243629.1 8 - 9 2519 74
XM_006243568.2 XP_006243630.1 9 - 10 2616 74
XM_006243569.2 XP_006243631.1 8 - 9 1330 74
XM_006243571.2 XP_006243633.1 8 - 9 1283 74
XM_006243572.2 XP_006243634.1 6 - 7 2151 74
XM_008766479.1 XP_008764701.1 8 - 9 774 74
XM_008766480.1 XP_008764702.1 8 - 9 770 74
XM_008766481.1 XP_008764703.1 8 - 9 774 74
XM_008766482.1 XP_008764704.1 12 - 13 1311 74
XM_008766483.1 XP_008764705.1 8 - 9 1097 74
XM_008766484.1 XP_008764706.1 12 - 13 1165 74
XM_008766485.1 XP_008764707.1 11 - 12 1069 74
XM_008766487.1 XP_008764709.1 9 - 10 900 74
XM_008766488.1 XP_008764710.1 6 - 7 610 74
GenBank mRNA BC168961.1 - 7 - 8 792 7444676 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800001331.rn.1

Set Membership:

Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
heart development
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transcription factor binding
protein domain specific binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17861 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 834717 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 163500 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 10915 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 120632 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 69500 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 668291 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 596922 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 278715 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 256779 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 613361 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 857821 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 571719 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 723230 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 260206 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 438061 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 150482 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 73064 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 429657 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 59677 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 77338 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 224589 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 125238 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 55123 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 268996 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 575455 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 13038 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 597089 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 252476 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 914027 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 554029 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 332737 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 139050 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 240727 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 759286 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 571250 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 364423 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 690120 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 570007 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 950442 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 225947 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 318827 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 27105 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 856984 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 908611 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 827722 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 420798 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 572308 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 602658 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 444858 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 693992 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 370812 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 278739 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 190984 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 100700 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 589808 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 285850 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 900670 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469502 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 123197 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 795001 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 929260 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 926331 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 503710 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 521231 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 771331 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 903409 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 673493 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 420174 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 535403 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 653139 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 955315 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 482550 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 654130 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 870387 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 574265 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 138344 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 786389 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 92126 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 163888 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 260613 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 709559 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 792814 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 45993 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 69977 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 874553 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 832838 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 274180 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 192022 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 263958 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 694587 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 614023 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743134 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 966026 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 482559 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 126635 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 51392 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 54273 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 34286 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 47053 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 108086 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 122076 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 42786 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 114337 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 125234 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7520 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 92880 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 1148 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 98280 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 173369 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 129215 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 141023 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 15305 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 149030 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17861 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17861 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17861 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17861 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20415 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 79353 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 7113 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 88772 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10912419 - -
Affymetrix Rat Gene 1.0 ST Array 10912413 - -
Affymetrix Rat Gene 1.0 ST Array 10912420 - -
Affymetrix Rat Gene 1.0 ST Array 10912417 - -
Affymetrix Rat Gene 1.0 ST Array 10912414 - -
Affymetrix Rat Gene 1.0 ST Array 10912421 - -
Affymetrix Rat Gene 1.0 ST Array 10912415 - -
Affymetrix Rat Gene 1.0 ST Array 10912418 - -
Affymetrix Rat Gene 1.0 ST Array 10912416 - -
Affymetrix Rat Gene 1.1 ST Array 10912412 - -
Rat Genome 230 2.0 Array 1379203_at - -
Rat Genome 230 2.0 Array 1379203_PM_at - -
Rat Genome 230 2.0 Array 1392405_PM_at - -
Rat Genome 230 2.0 Array 1392405_at - -
Affymetrix HT Rat Focus Array 1379203_PM_at - -
Affymetrix HT Rat Focus Array 1379203_at - -
Affymetrix Rat Clariom S Assay PSR0800009798.rn.1 TC0800001331.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800009797.rn.1 TC0800001331.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800009799.rn.1 TC0800001331.rn.1 RTA TC
Rat Expression Set 230 1392405_PM_at - -
Rat Expression Set 230 1379203_PM_at - -
Rat Expression Set 230 1392405_at - -
Rat Expression Set 230 1379203_at - -
Affymetrix Rat Gene 1.0 ST Array 10912412 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21667192 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1363133 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0