This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01404500_m1
See other RTP4 GE Assays ›
Gene Symbol:
Rtp4
Gene Name:
receptor (chemosensory) transporter protein 4
Gene Aliases:
RGD1306974
Chromosome Location:
Chr.11: 80638945 - 80650802 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01404500_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rtp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108321.1 NP_001101791.1 3 - 4 410 90
XM_006248521.2 XP_006248583.1 1 - 2 797 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100001867.rn.1

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s)

detection of chemical stimulus involved in sensory perception of bitter taste
protein targeting to membrane

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P736315 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 792924 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 716473 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 698140 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 184832 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 567919 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 99244 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 671734 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 788729 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 173203 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P736315 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P736315 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P736315 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P736315 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10755089 - -
Affymetrix Rat Gene 1.0 ST Array 10755090 - -
Affymetrix Rat Gene 1.1 ST Array 10755088 - -
Rat Genome 230 2.0 Array 1379285_at - -
Rat Genome 230 2.0 Array 1379285_PM_at - -
Affymetrix HT Rat Focus Array 1379285_PM_at - -
Affymetrix HT Rat Focus Array 1379285_at - -
Affymetrix Rat Clariom S Assay PSR1100011875.rn.1 TC1100001867.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100011882.rn.1 TC1100001867.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100011877.rn.1 TC1100001867.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100011883.rn.1 TC1100001867.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100011876.rn.1 TC1100001867.rn.1 RTA TC
Rat Expression Set 230 1379285_at - -
Rat Expression Set 230 1379285_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10755088 - -
Rat Genome U34 Set rc_AA819788_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21265598 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1368826 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline