This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01406436_m1
See other GNPDA1 GE Assays ›
Gene Symbol:
Gnpda1
Gene Name:
glucosamine-6-phosphate deaminase 1
Gene Aliases:
-
Chromosome Location:
Chr.18: 31479117 - 31489154 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01406436_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnpda1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134995.1 NP_001128467.1 1 - 2 272 126
GenBank mRNA BC169009.1 - 1 - 2 234 7312423 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800001313.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glucosamine catabolic process
N-acetylglucosamine metabolic process
generation of precursor metabolites and energy
glucosamine-6-phosphate deaminase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1800009432.rn.1 TC1800001313.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0