Assay Details
| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024741.1 | NP_001019912.1 | 6 - 7 | 695 | 63 | |
| XM_006248044.2 | XP_006248106.1 | 6 - 7 | 731 | 63 | ||
| GenBank mRNA | BC097460.1 | - | 6 - 7 | 695 | 7457454 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Chaf1b
Genomic Map