This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01407206_m1
See other IGSF5 GE Assays ›
Gene Symbol:
Igsf5
Gene Name:
immunoglobulin superfamily, member 5
Gene Aliases:
Jam4
Chromosome Location:
Chr.11: 36736579 - 36779614 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01407206_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006990.2 NP_001006991.1 7 - 8 1305 79
XM_006248155.2 XP_006248217.1 7 - 8 1170 79
XM_006248156.2 XP_006248218.1 6 - 7 869 79
GenBank mRNA AY439284.1 - 6 - 7 951 79
BC086342.1 - 7 - 8 1011 79
BC097947.1 - 7 - 8 1305 7373302 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100000358.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf5

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

single organismal cell-cell adhesion
regulation of neurogenesis
PDZ domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100001765.rn.1 TC1100000358.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100001767.rn.1 TC1100000358.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100001764.rn.1 TC1100000358.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100001766.rn.1 TC1100000358.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0