This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01407564_g1
See other TWF1 GE Assays ›
Gene Symbol:
Twf1
Gene Name:
twinfilin actin-binding protein 1
Gene Aliases:
Ptk9
Chromosome Location:
Chr.7: 135838647 - 135850542 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01407564_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008521.1 NP_001008521.1 8 - 9 892 69
GenBank mRNA BC086536.1 - 8 - 9 892 7106678 69

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Twf1

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-tyrosine phosphorylation
negative regulation of actin filament polymerization
sequestering of actin monomers
regulation of actin phosphorylation
barbed-end actin filament capping
actin monomer binding
protein tyrosine kinase activity
ATP binding
phosphatidylinositol-4,5-bisphosphate binding
protein complex binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P210120 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 165831 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 464334 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 725591 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 142742 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 902622 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 570669 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 113537 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 127744 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P210120 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P210120 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P210120 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P387290 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P558660 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P535434 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P210116 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P210120 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 26697 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 46989 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10906539 - -
Affymetrix Rat Gene 1.0 ST Array 10906542 - -
Affymetrix Rat Gene 1.0 ST Array 10906535 - -
Affymetrix Rat Gene 1.0 ST Array 10906541 - -
Affymetrix Rat Gene 1.0 ST Array 10906538 - -
Affymetrix Rat Gene 1.0 ST Array 10906540 - -
Affymetrix Rat Gene 1.0 ST Array 10906534 - -
Affymetrix Rat Gene 1.0 ST Array 10906537 - -
Affymetrix Rat Gene 1.0 ST Array 10906536 - -
Affymetrix Rat Gene 1.0 ST Array 10871694 - -
Affymetrix Rat Gene 1.1 ST Array 10906533 - -
Affymetrix Rat Gene 1.1 ST Array 10871693 - -
Rat Genome 230 2.0 Array 1371644_PM_at - -
Rat Genome 230 2.0 Array 1376365_PM_at - -
Rat Genome 230 2.0 Array 1371644_at - -
Rat Genome 230 2.0 Array 1376365_at - -
Affymetrix HT Rat Focus Array 1371644_at - -
Affymetrix HT Rat Focus Array 1376365_at - -
Affymetrix HT Rat Focus Array 1376365_PM_at - -
Affymetrix HT Rat Focus Array 1371644_PM_at - -
Affymetrix Rat Clariom S Assay PSR0700030081.rn.1 TC0700003764.rn.1 RTA TC
Rat Expression Set 230 1376365_PM_at - -
Rat Expression Set 230 1376365_at - -
Rat Expression Set 230 1371644_PM_at - -
Rat Expression Set 230 1371644_at - -
Affymetrix Rat Gene 1.0 ST Array 10871693 - -
Affymetrix Rat Gene 1.0 ST Array 10906533 - -
Rat Genome U34 Set rc_AA892851_g_at - -
Rat Genome U34 Set rc_AA892851_at - -
Rat Genome U34 Set rc_AI171354_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21406761 - -
ABI Rat Genome Survey Microarray 21907941 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1365408 - -
RatRef-12 Expression BeadChip ILMN_58628 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0