This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01408600_g1
See other POLR2E GE Assays ›
Gene Symbol:
Polr2e
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide E
Gene Aliases:
-
Chromosome Location:
Chr.7: 12532746 - 12536797 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01408600_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109614.1 NP_001103084.1 4 - 5 564 55
XM_006241009.2 XP_006241071.1 4 - 5 693 55
XM_006241010.1 XP_006241072.1 4 - 5 584 55
XM_006241011.1 XP_006241073.1 4 - 5 611 55
GenBank mRNA BC158787.1 - 4 - 5 522 7304442 55
FQ219797.1 - 4 - 5 496 55
FQ220392.1 - 4 - 5 494 55
FQ229401.1 - 4 - 5 611 55
FQ230290.1 - 4 - 5 499 55
FQ234550.1 - 4 - 5 544 55

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase I promoter
transcription from RNA polymerase II promoter
transcription from RNA polymerase III promoter
biological_process
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
molecular_function
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700002053.rn.1 TC0700000218.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0