This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01408628_g1
See other ATP5D GE Assays ›
Gene Symbol:
Atp5d
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit
Gene Aliases:
-
Chromosome Location:
Chr.7: 12426807 - 12432120 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01408628_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139106.1 NP_620806.1 1 - 2 190 91
XM_006240845.2 XP_006240907.1 2 - 3 705 91
GenBank mRNA BC161836.1 - 2 - 3 715 9098303 91
FQ214102.1 - 398 91
FQ223899.1 - 1 - 2 238 91
U00926.1 - 1 - 2 190 91

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5d

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
ATP synthesis coupled proton transport
ATP metabolic process
ATPase activity
protein complex binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700017287.rn.1 TC0700002176.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700017289.rn.1 TC0700002176.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700017292.rn.1 TC0700002176.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700017288.rn.1 TC0700002176.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700017291.rn.1 TC0700002176.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700017293.rn.1 TC0700002176.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700017290.rn.1 TC0700002176.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0