This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01409509_g1
See other ATP5J2 GE Assays ›
Gene Symbol:
Atp5j2
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F2
Gene Aliases:
-
Chromosome Location:
Chr.12: 11191799 - 11198369 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01409509_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5j2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271117.1 NP_001258046.1 2 - 3 164 71
GenBank mRNA FQ210117.1 - 2 - 3 151 71
FQ211282.1 - 2 - 3 151 71
FQ216873.1 - 2 - 3 139 71
FQ217338.1 - 2 - 3 138 71
FQ217676.1 - 2 - 3 139 71
FQ218321.1 - 2 - 3 153 71
FQ221351.1 - 2 - 3 143 71
FQ221691.1 - 2 - 3 151 71
FQ221732.1 - 2 - 3 133 71
FQ222492.1 - 2 - 3 138 71
FQ223295.1 - 2 - 3 141 71
FQ223575.1 - 2 - 3 136 71
FQ223738.1 - 2 - 3 139 71
FQ223886.1 - 1 - 2 117 71
FQ223939.1 - 2 - 3 139 71
FQ224192.1 - 2 - 3 139 71
FQ224234.1 - 2 - 3 153 71
FQ224423.1 - 2 - 3 151 71
FQ224479.1 - 2 - 3 151 71
FQ224557.1 - 2 - 3 151 71
FQ224602.1 - 2 - 3 152 71
FQ228438.1 - 2 - 3 139 71

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
proton transport
ATP metabolic process
ATPase activity

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038960 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038963 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038963 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038960 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038963 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038960 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT Rat Focus Array 1371398_PM_at - -
Affymetrix HT Rat Focus Array 1371398_at - -
Affymetrix Rat Clariom S Assay PSR1200001282.rn.1 TC1200000188.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200001283.rn.1 TC1200000188.rn.1 RTA TC
Rat Genome U34 Set rc_AI103097_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0