This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01409511_g1
See other ATP5J2 GE Assays ›
Gene Symbol:
Atp5j2
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F2
Gene Aliases:
-
Chromosome Location:
Chr.12: 11191799 - 11198369 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01409511_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5j2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271117.1 NP_001258046.1 1 - 2 51 62
GenBank mRNA FQ210117.1 - 1 - 2 38 62
FQ211282.1 - 1 - 2 38 62
FQ218321.1 - 1 - 2 40 62
FQ221691.1 - 1 - 2 38 62
FQ224234.1 - 1 - 2 40 62
FQ224423.1 - 1 - 2 38 62
FQ224479.1 - 1 - 2 38 62
FQ224557.1 - 1 - 2 38 62
FQ224602.1 - 1 - 2 39 62
FQ224807.1 - 1 - 2 38 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001271117.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
proton transport
ATP metabolic process
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200001280.rn.1 TC1200000188.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200001278.rn.1 TC1200000188.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0