Assay Details
| Gene Symbol: Disp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105983.1 | NP_001099453.1 | 5 - 6 | 977 | 58 | |
| XM_006250381.2 | XP_006250443.1 | 4 - 5 | 846 | 58 | ||
| XM_006250383.2 | XP_006250445.1 | 3 - 4 | 1307 | 58 | ||
| XM_006250384.2 | XP_006250446.1 | 3 - 4 | 786 | 58 | ||
| XM_008769827.1 | XP_008768049.1 | 6 - 7 | 1671 | 58 | ||
| XM_008769828.1 | XP_008768050.1 | 4 - 5 | 957 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
smoothened signaling pathway
patched ligand maturation
determination of left/right symmetry
pattern specification process
biological_process
embryonic pattern specification
dorsal/ventral pattern formation
peptide transport
diaphragm development
molecular_function
hedgehog receptor activity
peptide transporter activity
smoothened signaling pathway
patched ligand maturation
determination of left/right symmetry
pattern specification process
biological_process
embryonic pattern specification
dorsal/ventral pattern formation
peptide transport
diaphragm development
molecular_function
hedgehog receptor activity
peptide transporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P002949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44071 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002949 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002949 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P002949 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P808456 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71116 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10770475 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770478 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770473 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770474 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770472 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770476 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770477 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10770471 | - | - |
| Rat Genome 230 2.0 Array | 1374219_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374219_at | - | - |
| Affymetrix HT Rat Focus Array | 1374219_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374219_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300015113.rn.1 | TC1300002116.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300015114.rn.1 | TC1300002116.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300015115.rn.1 | TC1300002116.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374219_PM_at | - | - |
| Rat Expression Set 230 | 1374219_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770471 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21204703 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369055 | - | - |

Disp1
Genomic Map