This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01412066_m1
See other FH GE Assays ›
Gene Symbol:
Fh
Gene Name:
fumarate hydratase
Gene Aliases:
Fh1
Chromosome Location:
Chr.13: 93651486 - 93677371 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01412066_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017005.2 NP_058701.2 1 - 2 139 122
GenBank mRNA BC087598.1 - 1 - 2 139 7311498 122
J04473.1 - 1 - 2 150 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300001997.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fh

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
fumarate metabolic process
malate metabolic process
homeostasis of number of cells within a tissue
fumarate hydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300014280.rn.1 TC1300001997.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300014281.rn.1 TC1300001997.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0