This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01412109_g1
See other SDHC GE Assays ›
Gene Symbol:
Sdhc
Gene Name:
succinate dehydrogenase complex, subunit C, integral membrane protein
Gene Aliases:
-
Chromosome Location:
Chr.13: 89498047 - 89518939 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01412109_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdhc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005534.1 NP_001005534.1 4 - 5 244 103
XM_008769731.1 XP_008767953.1 5 - 6 291 103
GenBank mRNA BC082027.1 - 4 - 5 244 7130052 103
FQ210665.1 - 4 - 5 246 103
FQ213406.1 - 4 - 5 263 103
FQ215668.1 - 5 - 6 298 103
FQ215689.1 - 4 - 5 246 103
FQ216122.1 - 4 - 5 262 103
FQ216612.1 - 4 - 5 263 103
FQ216658.1 - 4 - 5 246 103
FQ217679.1 - 4 - 5 263 103
FQ218940.1 - 4 - 5 263 103
FQ224704.1 - 4 - 5 246 103
FQ225195.1 - 4 - 5 257 103
FQ229651.1 - 4 - 5 263 103
FQ231805.1 - 4 - 5 263 103

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sdhc

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
biological_process
succinate dehydrogenase activity
molecular_function
electron carrier activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300013499.rn.1 TC1300001901.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0