This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01412766_m1
See other RAC1 GE Assays ›
Gene Symbol:
Rac1
Gene Name:
ras-related C3 botulinum toxin substrate 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 13090338 - 13111835 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01412766_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rac1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134366.1 NP_599193.1 4 - 5 289 86
XM_008768986.1 XP_008767208.1 5 - 6 380 86
GenBank mRNA AY491395.1 - 4 - 5 289 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000223.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rac1

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
auditory receptor cell morphogenesis
mast cell chemotaxis
epithelial cell morphogenesis
endocytosis
phagocytosis, engulfment
chemotaxis
hyperosmotic response
cytoskeleton organization
actin filament organization
cell adhesion
G-protein coupled receptor signaling pathway
small GTPase mediated signal transduction
axon guidance
metabolic process
cell proliferation
positive regulation of lamellipodium assembly
positive regulation of cell-substrate adhesion
regulation of neuron maturation
dendrite development
cell migration
Rac protein signal transduction
cerebral cortex radially oriented cell migration
embryonic olfactory bulb interneuron precursor migration
cerebral cortex GABAergic interneuron development
regulation of cell morphogenesis
lamellipodium assembly
actin cytoskeleton organization
actin filament polymerization
regulation of cell migration
positive regulation of actin filament polymerization
ruffle organization
negative regulation of interleukin-23 production
substrate adhesion-dependent cell spreading
positive regulation of phosphatidylinositol 3-kinase activity
engulfment of apoptotic cell
cell-cell junction organization
bone resorption
positive regulation of DNA replication
anatomical structure arrangement
dendrite morphogenesis
cell motility
homeostasis of number of cells within a tissue
positive regulation of stress fiber assembly
localization within membrane
positive regulation of focal adhesion assembly
synaptic transmission, GABAergic
Wnt signaling pathway, planar cell polarity pathway
regulation of respiratory burst
cellular response to mechanical stimulus
semaphorin-plexin signaling pathway
dopaminergic neuron differentiation
protein localization to plasma membrane
positive regulation of neutrophil chemotaxis
cochlea morphogenesis
ruffle assembly
positive regulation of substrate adhesion-dependent cell spreading
GTPase activity
protein binding
GTP binding
Rab GTPase binding
enzyme binding
protein kinase binding
GTP-dependent protein binding
thioesterase binding
histone deacetylase binding
Rho GDP-dissociation inhibitor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200001602.rn.1 TC1200000223.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0