This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01412873_m1
See other NUDT1 GE Assays ›
Gene Symbol:
Nudt1
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 1
Gene Aliases:
Mth1
Chromosome Location:
Chr.12: 16391578 - 16398746 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01412873_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_057120.1 NP_476461.1 1 - 2 163 85
XM_006248911.2 XP_006248973.1 2 - 3 214 85
GenBank mRNA D49977.1 - 1 - 2 163 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200001179.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt1

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide catabolic process
dGTP catabolic process
DNA repair
DNA protection
dATP catabolic process
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
snoRNA binding
8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity
GTP diphosphatase activity
metal ion binding
ATP diphosphatase activity
m7G(5')pppN diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200010051.rn.1 TC1200001179.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200010052.rn.1 TC1200001179.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0