Assay Details
| Gene Symbol: Gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134756.1 | NP_001128228.1 | 2 - 3 | 234 | 73 | |
| NM_001134757.1 | NP_001128229.1 | 1 - 2 | 240 | 73 | ||
| XM_006251782.2 | XP_006251844.1 | 2 - 3 | 251 | 73 | ||
| XM_006251783.2 | XP_006251845.1 | 2 - 3 | 299 | 73 | ||
| XM_006251784.2 | XP_006251846.1 | 2 - 3 | 247 | 73 | ||
| GenBank mRNA | BC160861.1 | - | 1 - 2 | 240 | 7322586 | 73 |
| CA339205.1 | - | 2 - 3 | 304 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gnpnat1
Genomic Map