This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01415035_m1
See other HSD11B1 GE Assays ›
Gene Symbol:
Hsd11b1
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases:
LRRGT00065
Chromosome Location:
Chr.13: 111946626 - 111996531 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01415035_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd11b1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017080.2 NP_058776.2 2 - 3 545 65
XM_006250471.2 XP_006250533.1 3 - 4 326 65
XM_006250473.2 XP_006250535.1 3 - 4 361 65
GenBank mRNA AF542063.1 - 1 - 2 145 65
BC078865.1 - 2 - 3 366 7109256 65
FQ209646.1 - 2 - 3 312 65
FQ209829.1 - 2 - 3 312 65
FQ210302.1 - 2 - 3 310 65
FQ210563.1 - 2 - 3 310 65
FQ210596.1 - 2 - 3 312 65
FQ211059.1 - 2 - 3 309 65
FQ211404.1 - 2 - 3 311 65
FQ214397.1 - 3 - 4 368 65
FQ218558.1 - 2 - 3 310 65
FQ218584.1 - 2 - 3 312 65
FQ219123.1 - 2 - 3 312 65
FQ219241.1 - 2 - 3 311 65
FQ219385.1 - 2 - 3 310 65
FQ219394.1 - 2 - 3 310 65
FQ219517.1 - 2 - 3 312 65
FQ219567.1 - 2 - 3 321 65
FQ219624.1 - 2 - 3 245 65
FQ220122.1 - 2 - 3 310 65
J05107.1 - 2 - 3 293 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300002245.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsd11b1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glucocorticoid biosynthetic process
glucocorticoid catabolic process
metabolic process
phthalate metabolic process
lung development
response to nutrient levels
regulation of pentose-phosphate shunt
oxidation-reduction process
cellular response to estradiol stimulus
11-beta-hydroxysteroid dehydrogenase [NAD(P)] activity
steroid binding
oxidoreductase activity
NADP binding
11-beta-hydroxysteroid dehydrogenase (NADP+) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0