This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01415059_m1
See other RPS6KC1 GE Assays ›
Gene Symbol:
Rps6kc1
Gene Name:
ribosomal protein S6 kinase, polypeptide C1
Gene Aliases:
-
Chromosome Location:
Chr.13: 109338761 - 109483924 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01415059_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps6kc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105984.1 NP_001099454.1 9 - 10 1196 117
XM_006250459.2 XP_006250521.1 10 - 11 1375 117
XM_006250460.2 XP_006250522.1 9 - 10 1282 117
XM_006250461.2 XP_006250523.1 10 - 11 1514 117
XM_008769829.1 XP_008768051.1 10 - 11 1375 117
XM_008769830.1 XP_008768052.1 9 - 10 1015 117
XM_008769831.1 XP_008768053.1 9 - 10 1152 117
XM_008769832.1 XP_008768054.1 3 - 4 3726 117
XM_008769833.1 XP_008768055.1 3 - 4 411 117
XM_008769834.1 XP_008768056.1 4 - 5 345 117
GenBank mRNA BC064438.1 - 10 - 11 1446 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300002202.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein kinase activity
ATP binding
phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300015658.rn.1 TC1300002202.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline