This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01416008_m1
See other POLR3K GE Assays ›
Gene Symbol:
Polr3k
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide K
Gene Aliases:
-
Chromosome Location:
Chr.3: 177374812 - 177379009 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01416008_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3k
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014259.1 NP_001014281.1 1 - 2 176 76
GenBank mRNA BC089990.1 - 1 - 2 176 7319381 76
FQ212572.1 - 1 - 2 198 76
FQ213710.1 - 1 - 2 199 76
FQ213830.1 - 1 - 2 210 76
FQ216547.1 - 1 - 2 199 76
FQ226947.1 - 1 - 2 181 76
FQ232144.1 - 1 - 2 210 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300002614.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr3k

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

termination of RNA polymerase III transcription
biological_process
RNA polymerase III activity
molecular_function
DNA binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300019418.rn.1 TC0300002614.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300019417.rn.1 TC0300002614.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0