This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01416465_m1
See other GPX7 GE Assays ›
Gene Symbol:
Gpx7
Gene Name:
glutathione peroxidase 7
Gene Aliases:
-
Chromosome Location:
Chr.5: 128001766 - 128009610 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01416465_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpx7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106673.1 NP_001100143.1 2 - 3 455 57
GenBank mRNA FQ229888.1 - 2 - 3 400 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500003326.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
oxidation-reduction process
peroxidase activity
glutathione peroxidase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100250 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 240430 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 648165 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 41800 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 809039 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 374453 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100250 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100250 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100250 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1038773 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10878637 - -
Affymetrix Rat Gene 1.0 ST Array 10878636 - -
Affymetrix Rat Gene 1.0 ST Array 10878639 - -
Affymetrix Rat Gene 1.0 ST Array 10878638 - -
Affymetrix Rat Gene 1.1 ST Array 10878635 - -
Rat Genome 230 2.0 Array 1372125_at - -
Rat Genome 230 2.0 Array 1372125_PM_at - -
Affymetrix HT Rat Focus Array 1372125_at - -
Affymetrix HT Rat Focus Array 1372125_PM_at - -
Affymetrix Rat Clariom S Assay PSR0500024212.rn.1 TC0500003326.rn.1 RTA TC
Rat Expression Set 230 1372125_at - -
Rat Expression Set 230 1372125_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10878635 - -
Rat Genome U34 Set rc_AA893022_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20705604 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1373821 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7kcm7:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0