This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01416610_m1
See other TAL1 GE Assays ›
Gene Symbol:
Tal1
Gene Name:
T-cell acute lymphocytic leukemia 1
Gene Aliases:
Tal-1
Chromosome Location:
Chr.5: 133864403 - 133878604 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01416610_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tal1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107958.2 NP_001101428.2 3 - 4 870 100
XM_006238615.2 XP_006238677.1 3 - 4 736 100
XM_006238617.1 XP_006238679.1 3 - 4 944 100
XM_006238618.2 XP_006238680.1 2 - 3 588 100
XM_008763967.1 XP_008762189.1 3 - 4 1255 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500001235.rn.1

Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
angiogenesis
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
locomotory behavior
biological_process
spinal cord association neuron differentiation
hemopoiesis
myeloid cell differentiation
neuron differentiation
erythrocyte differentiation
megakaryocyte differentiation
platelet formation
basophil differentiation
positive regulation of protein complex assembly
embryonic hemopoiesis
megakaryocyte development
regulation of cell proliferation
erythrocyte maturation
cell fate commitment
regulation of myeloid cell differentiation
positive regulation of erythrocyte differentiation
positive regulation of chromatin assembly or disassembly
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
generation of neurons
positive regulation of cell division
astrocyte fate commitment
definitive hemopoiesis
hemangioblast cell differentiation
hematopoietic stem cell differentiation
regulation of mast cell differentiation
regulation of stem cell population maintenance
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
molecular_function
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
enzyme binding
histone deacetylase binding
transcription regulatory region DNA binding
protein heterodimerization activity
E-box binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500008414.rn.1 TC0500001235.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500008416.rn.1 TC0500001235.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500008409.rn.1 TC0500001235.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500008415.rn.1 TC0500001235.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500008408.rn.1 TC0500001235.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500008403.rn.1 TC0500001235.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0