This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01417968_m1
See other GLCE GE Assays ›
Gene Symbol:
Glce
Gene Name:
glucuronic acid epimerase
Gene Aliases:
RGD1565253
Chromosome Location:
Chr.8: 67006588 - 67073714 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01417968_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glce
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_343404.6 XP_343405.2 1 - 2 202 178

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_343404.6 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
racemase and epimerase activity, acting on carbohydrates and derivatives
heparosan-N-sulfate-glucuronate 5-epimerase activity
UDP-glucuronate 5'-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline