This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01419129_m1
See other DGAT2L6 GE Assays ›
Gene Symbol:
Dgat2l6
Gene Name:
diacylglycerol O-acyltransferase 2-like 6
Gene Aliases:
-
Chromosome Location:
Chr.X: 70354357 - 70377887 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01419129_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgat2l6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109370.1 NP_001102840.1 4 - 5 560 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0X00000598.rn.1

Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
transferase activity, transferring acyl groups other than amino-acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00003657.rn.1 TC0X00000598.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0