This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01421463_m1
See other NXPE2 GE Assays ›
Gene Symbol:
Nxpe2
Gene Name:
neurexophilin and PC-esterase domain family, member 2
Gene Aliases:
-
Chromosome Location:
Chr.8: 60333201 - 60359944 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01421463_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nxpe2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002729901.4 XP_002729947.2 4 - 5 1631 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102265 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028838 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P028838 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102265 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P102265 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P028838 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21678948 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0