This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01421562_m1
See other HACD3 GE Assays ›
Gene Symbol:
Hacd3
Gene Name:
3-hydroxyacyl-CoA dehydratase 3
Gene Aliases:
Ptplad1, RGD1565496
Chromosome Location:
Chr.8: 70471572 - 70509874 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01421562_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hacd3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106831.1 NP_001100301.1 8 - 9 978 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800002772.rn.1

Set Membership:

Amplicon length between 71 and 85 Protein Phosphatase Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

I-kappaB kinase/NF-kappaB signaling
JNK cascade
Rho protein signal transduction
fatty acid elongation
very long-chain fatty acid biosynthetic process
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
3-hydroxyacyl-CoA dehydratase activity
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800022516.rn.1 TC0800002772.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0