This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01422108_m1
See other PIF1 GE Assays ›
Gene Symbol:
Pif1
Gene Name:
PIF1 5'-to-3' DNA helicase
Gene Aliases:
-
Chromosome Location:
Chr.8: 71124477 - 71133460 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01422108_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pif1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044253.1 NP_001037718.1 6 - 7 1180 71
XM_006243271.2 XP_006243333.1 6 - 7 2117 71
XM_008766324.1 XP_008764546.1 7 - 8 1836 71
XM_008766325.1 XP_008764547.1 7 - 8 1808 71
XM_008766326.1 XP_008764548.1 7 - 8 1767 71
XM_008766327.1 XP_008764549.1 7 - 8 1739 71
GenBank mRNA DQ489562.1 - 6 - 7 1180 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000945.rn.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial genome maintenance
telomere maintenance
DNA repair
DNA recombination
regulation of telomere maintenance
negative regulation of telomere maintenance via telomerase
DNA duplex unwinding
negative regulation of telomerase activity
magnesium ion binding
ATP binding
telomerase inhibitor activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity
ATP-dependent 5'-3' DNA/RNA helicase activity
telomeric DNA binding
ATP-dependent 5'-3' DNA helicase activity
G-quadruplex DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800006866.rn.1 TC0800000945.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0