This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01422465_g1
See other RBPMS2 GE Assays ›
Gene Symbol:
Rbpms2
Gene Name:
RNA binding protein with multiple splicing 2
Gene Aliases:
RGD1561222
Chromosome Location:
Chr.8: 71165373 - 71200299 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01422465_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpms2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001173426.1 NP_001166897.1 3 - 4 309 102
XM_006243274.2 XP_006243336.1 3 - 4 1995 102
XM_008766329.1 XP_008764551.1 2 - 3 418 102
GenBank mRNA BC169038.1 - 3 - 4 309 9090232 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000947.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Process(es)

nucleotide binding
nucleic acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800006885.rn.1 TC0800000947.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800006880.rn.1 TC0800000947.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800006886.rn.1 TC0800000947.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline